siRNA Details
| Gene Symbol: BMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021073.2 | NP_066551.1 | 1 | 1086 | ||
| XM_005249304.3 | XP_005249361.1 | 1 | 1134 | |||
| XM_011514816.2 | XP_011513118.1 | 1 | 382 | |||
| XM_011514817.2 | XP_011513119.1 | 1 | 1134 | |||
| GenBank mRNA | AK291559.1 | - | 1 | 1099 | ||
| AK303576.1 | - | 1 | 388 | |||
| BC027958.1 | - | 1 | 1086 | 5209637 | ||
| KC485577.1 | - | 1 | 374 | |||
| M60314.1 | - | 1 | 1072 | |||

BMP5
Genomic Map