siRNA Details
| Gene Symbol: CBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005188.3 | NP_005179.2 | 16 | 2761 | ||
| XM_017018511.1 | XP_016874000.1 | 15 | 2196 | |||
| GenBank mRNA | BC132733.1 | - | 16 | 2690 | 40146755 | |
| BC136463.1 | - | 16 | 2690 | 9020452 | ||
| X57110.1 | - | 16 | 2767 | |||

CBL
Genomic Map