siRNA Details
| Gene Symbol: CBSL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321073.1 | NP_001308002.1 | 17 | 1863 | ||
| XM_011546094.1 | XP_011544396.1 | 18 | 1797 | |||
| XM_011546095.2 | XP_011544397.1 | 18 | 1939 | |||
| XM_011546097.2 | XP_011544399.1 | 19 | 1966 | |||
| XM_011546098.1 | XP_011544400.1 | 18 | 2304 | |||
| XM_011546099.1 | XP_011544401.1 | 17 | 1755 | |||
| XM_017028209.1 | XP_016883698.1 | 15 | 1960 | |||
| XM_017028210.1 | XP_016883699.1 | 15 | 1960 | |||
| XM_017028211.1 | XP_016883700.1 | 19 | 2302 | |||
| XM_017028212.1 | XP_016883701.1 | 20 | 2358 | |||
| XM_017028213.1 | XP_016883702.1 | 18 | 1797 | |||
| XM_017028214.1 | XP_016883703.1 | 18 | 2257 | |||
| XM_017028215.1 | XP_016883704.1 | 18 | 1872 | |||
| XM_017028216.1 | XP_016883705.1 | 17 | 2262 | |||
| XM_017028217.1 | XP_016883706.1 | 17 | 1755 | |||
| XM_017028218.1 | XP_016883707.1 | 18 | 1924 | |||
| GenBank mRNA | BC007257.1 | - | 17 | 1833 | 3028099 | |

CBS
Genomic Map