siRNA Details
| Gene Symbol: CCNF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323538.1 | NP_001310467.1 | 4 | 428 | ||
| NM_001761.2 | NP_001752.2 | 5 | 532 | |||
| XM_017023822.1 | XP_016879311.1 | 5 | 692 | |||
| XM_017023823.1 | XP_016879312.1 | 5 | 692 | |||
| GenBank mRNA | AB208829.1 | - | 4 | 339 | ||
| AK313371.1 | - | 5 | 485 | |||
| BC012349.1 | - | 5 | 456 | 4634116 | ||
| U17105.1 | - | 5 | 457 | |||
| Z36714.1 | - | 5 | 487 | |||

CCNF
Genomic Map