siRNA Details
| Gene Symbol: CD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK223393.1 | - | 4 | 695 | ||
| AK301154.1 | - | 4 | 718 | |||
| AK303367.1 | - | 4 | 519 | |||
| AK310676.1 | - | 4 | 557 | |||
| AK313614.1 | - | 4 | 724 | |||
| BC033583.1 | - | 4 | 679 | 5227138 | ||
| CR996443.1 | - | 4 | 779 | |||
| M14362.1 | - | 4 | 675 | |||
| M16336.1 | - | 4 | 689 | |||
| M16445.1 | - | 4 | 672 | |||

CD2
Genomic Map