siRNA Details
| Gene Symbol: CD3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000733.3 | NP_000724.1 | 2, 3 | 299 | ||
| GenBank mRNA | AK292612.1 | - | 2, 3 | 148 | ||
| AK311641.1 | - | 2, 3 | 149 | |||
| BI910359.1 | - | 2, 3 | 334 | |||
| BP366535.1 | - | 2, 3 | 316 | |||
| X03884.1 | - | 1, 2 | 97 | |||

CD3E
Genomic Map