siRNA Details
| Gene Symbol: CD3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000073.2 | NP_000064.1 | 1 | 120 | ||
| XM_005271724.3 | XP_005271781.1 | 1 | 157 | |||
| XM_006718941.2 | XP_006719004.1 | 1 | 108 | |||
| GenBank mRNA | AK223314.1 | - | 1 | 73 | ||
| AK299997.1 | - | 1 | 120 | |||
| AK310578.1 | - | 1 | 68 | |||
| AK310622.1 | - | 1 | 100 | |||
| AK313966.1 | - | 1 | 108 | |||
| BC113830.1 | - | 1 | 66 | 40029587 | ||
| DC378393.1 | - | 1 | 120 | |||
| X04145.1 | - | 1 | 77 | |||
| X60492.1 | - | 1 | 77 | |||

CD3G
Genomic Map