siRNA Details
| Gene Symbol: CD5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005894.2 | NP_005885.1 | 1, 2 | 182 | ||
| XM_005245602.1 | XP_005245659.1 | 1, 2 | 182 | |||
| XM_017002806.1 | XP_016858295.1 | 1, 2 | 182 | |||
| GenBank mRNA | AF011429.1 | - | 1, 2 | 102 | ||
| AK292040.1 | - | 1, 2 | 103 | |||
| AY358494.1 | - | 1, 2 | 136 | |||
| BC033586.1 | - | 1, 2 | 70 | 5225975 | ||
| DA957550.1 | - | 1, 2 | 182 | |||
| U82812.1 | - | 1, 2 | 99 | |||

CD5L
Genomic Map