siRNA Details
| Gene Symbol: CD27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001242.4 | NP_001233.1 | 2, 3 | 476 | ||
| XM_017020232.1 | XP_016875721.1 | 2, 3 | 494 | |||
| XM_017020234.1 | XP_016875723.1 | 2, 3 | 494 | |||
| GenBank mRNA | AK315732.1 | - | 2, 3 | 495 | ||
| BC012160.1 | - | 2, 3 | 462 | 4575359 | ||
| M63928.1 | - | 2, 3 | 365 | |||

CD27
Genomic Map