siRNA Details
| Gene Symbol: TNFSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001244.3 | NP_001235.1 | 4 | 756 | ||
| GenBank mRNA | AK313619.1 | - | 4 | 756 | ||
| AL583443.3 | - | 4 | 660 | |||
| BC093630.1 | - | 4 | 538 | 7939475 | ||
| BC111939.1 | - | 4 | 538 | 8327407 | ||
| L09753.1 | - | 4 | 578 | |||

TNFSF8
Genomic Map