siRNA Details
| Gene Symbol: CD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000560.3 | NP_000551.1 | 7 | 690 | ||
| NM_001040033.1 | NP_001035122.1 | 8 | 695 | |||
| NM_001320638.1 | NP_001307567.1 | 5 | 513 | |||
| GenBank mRNA | AK298723.1 | - | 5 | 460 | ||
| AK313582.1 | - | 7 | 643 | |||
| BC035456.1 | - | 8 | 677 | |||
| BC040693.1 | - | 7 | 620 | 5498854 | ||
| BT019643.1 | - | 6 | 524 | |||
| M37033.1 | - | 7 | 617 | |||
| M60871.1 | - | 7 | 597 | |||

CD53
Genomic Map