siRNA Details
| Gene Symbol: CD63 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001257389.1 | NP_001244318.1 | 3 | 635 | ||
| NM_001257390.1 | NP_001244319.1 | 3 | 378 | |||
| NM_001257391.1 | NP_001244320.1 | 2 | 373 | |||
| NM_001257392.1 | NP_001244321.1 | 2 | 174 | |||
| NM_001257400.1 | NP_001244329.1 | 2 | 203 | |||
| NM_001257401.1 | NP_001244330.1 | 2 | 343 | |||
| NM_001267698.1 | NP_001254627.1 | 3 | 425 | |||
| NM_001780.5 | NP_001771.1 | 3 | 429 | |||
| GenBank mRNA | AF508304.1 | - | 2 | 171 | ||
| AK311893.1 | - | 3 | 300 | |||
| AL582859.3 | - | 2 | 248 | |||
| BC002349.2 | - | 3 | 317 | 2819554 | ||
| BC013017.2 | - | 3 | 273 | 3010594 | ||
| BE407235.1 | - | 2 | 372 | |||
| BG478842.1 | - | 3 | 379 | |||
| BG766593.1 | - | 2 | 202 | |||
| BQ434569.1 | - | 344 | ||||
| BT007073.1 | - | 2 | 232 | |||
| BT020137.1 | - | 2 | 232 | |||
| BT020138.1 | - | 2 | 232 | |||
| BU167765.1 | - | 3 | 435 | |||
| BU190521.1 | - | 2 | 368 | |||
| CD250685.1 | - | 3 | 488 | |||
| CN296813.1 | - | 3 | 392 | |||
| CN296834.1 | - | 3 | 417 | |||
| CN484590.1 | - | 2 | 243 | |||
| CR542096.1 | - | 2 | 232 | |||
| DB495090.1 | - | 3 | 460 | |||
| EB386351.1 | - | 3 | 364 | |||
| KF998086.1 | - | 2 | 232 | |||
| M58485.1 | - | 3 | 287 | |||
| M59907.1 | - | 3 | 282 | |||
| S93788.1 | - | 2 | 256 | |||
| X07982.1 | - | 3 | 259 | |||

CD63
Genomic Map