siRNA Details
| Gene Symbol: CDC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253.3 | NP_001244.1 | 14 | 2350 | ||
| GenBank mRNA | AB007892.2 | - | 14 | 2122 | ||
| AK128737.1 | - | 14 | 2158 | |||
| AK299738.1 | - | 14 | 2077 | |||
| BC001568.1 | - | 14 | 2139 | 3452352 | ||
| U86753.1 | - | 14 | 2040 | |||

CDC5L
Genomic Map