siRNA Details
| Gene Symbol: CDH12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317227.1 | NP_001304156.1 | 12 | 2661 | ||
| NM_001317228.1 | NP_001304157.1 | 12 | 2656 | |||
| NM_004061.4 | NP_004052.2 | 15 | 3290 | |||
| XM_011513927.2 | XP_011512229.1 | 13 | 2567 | |||
| XM_017008920.1 | XP_016864409.1 | 14 | 2917 | |||
| XM_017008921.1 | XP_016864410.1 | 15 | 3010 | |||
| GenBank mRNA | AK092176.1 | - | 10 | 1940 | ||
| AK295123.1 | - | 12 | 2656 | |||
| AK314800.1 | - | 12 | 2661 | |||
| BC047608.1 | - | 15 | 2995 | 5751948 | ||
| L33477.1 | - | 15 | 3291 | |||

CDH12
Genomic Map