siRNA Details
| Gene Symbol: CDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004065.2 | NP_004056.2 | 1 | 311 | ||
| GenBank mRNA | BC113472.1 | - | 1 | 236 | 8322524 | |
| BC113474.1 | - | 1 | 236 | 8322526 | ||
| M16965.1 | - | 1 | 179 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDR1| Gene Symbol: CDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004065.2 | NP_004056.2 | 1 | 311 | ||
| GenBank mRNA | BC113472.1 | - | 1 | 236 | 8322524 | |
| BC113474.1 | - | 1 | 236 | 8322526 | ||
| M16965.1 | - | 1 | 179 | |||