siRNA Details
Gene Symbol: CDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005193.1 | NP_005184.1 | 1 | 174 | ||
GenBank mRNA | BC128233.1 | - | 1 | 174 | 40114658 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: CDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005193.1 | NP_005184.1 | 1 | 174 | ||
GenBank mRNA | BC128233.1 | - | 1 | 174 | 40114658 |