siRNA Details
| Gene Symbol: CLGN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130675.1 | NP_001124147.1 | 8 | 770 | ||
| NM_004362.2 | NP_004353.1 | 7 | 686 | |||
| GenBank mRNA | AK093096.1 | - | 7 | 1249 | ||
| AK290819.1 | - | 7 | 683 | |||
| AK299243.1 | - | 6 | 475 | |||
| BC028357.2 | - | 7 | 640 | 4830135 | ||
| D86322.1 | - | 7 | 653 | |||

CLGN
Genomic Map