siRNA Details
| Gene Symbol: CEACAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291484.2 | NP_001278413.1 | 5 | 1215 | ||
| NM_001308398.1 | NP_001295327.1 | 5 | 1212 | |||
| NM_004363.5 | NP_004354.3 | 5 | 1215 | |||
| XM_005258413.3 | XP_005258470.2 | 5 | 1184 | |||
| XM_017026145.1 | XP_016881634.1 | 5 | 1181 | |||
| XM_017026146.1 | XP_016881635.1 | 4 | 929 | |||
| GenBank mRNA | AB852566.1 | - | 3 | 534 | ||
| AK223101.1 | - | 5 | 1151 | |||
| BC034671.1 | - | 5 | 1163 | 5184800 | ||
| M15042.1 | - | 5 | 1057 | |||
| M16234.1 | - | 1 | 79 | |||
| M17191.1 | - | 1 | 125 | |||
| M29540.1 | - | 5 | 1182 | |||
| X16455.1 | - | 5 | 1054 | |||

CEACAM5
Genomic Map