siRNA Details
| Gene Symbol: CENPF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016343.3 | NP_057427.3 | 12 | 4009 | ||
| XM_011509082.2 | XP_011507384.1 | 12 | 4017 | |||
| XM_017000086.1 | XP_016855575.1 | 12 | 3996 | |||
| GenBank mRNA | U19769.1 | - | 4005 | |||
| U30872.1 | - | 12 | 3904 | |||

CENPF
Genomic Map