siRNA Details
| Gene Symbol: CHGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001819.2 | NP_001810.2 | 4 | 1495 | ||
| GenBank mRNA | AB209712.1 | - | 4 | 1292 | ||
| AK225125.1 | - | 4 | 1313 | |||
| AK289386.1 | - | 4 | 1315 | |||
| AK293536.1 | - | 4 | 1049 | |||
| AK312237.1 | - | 4 | 1315 | |||
| BC000375.2 | - | 4 | 1293 | 2820258 | ||
| CR456726.1 | - | 4 | 1189 | |||
| Y00064.1 | - | 4 | 1301 | |||

CHGB
Genomic Map