siRNA Details
| Gene Symbol: CLCN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256944.1 | NP_001243873.1 | 7 | 1114 | ||
| NM_001830.3 | NP_001821.2 | 9 | 1369 | |||
| GenBank mRNA | AB019432.1 | - | 8 | 1214 | ||
| AF170492.1 | - | 7 | 939 | |||
| AK225287.1 | - | 9 | 1330 | |||
| AK289564.1 | - | 9 | 1316 | |||
| AK299611.1 | - | 6 | 747 | |||
| BC036068.1 | - | 8 | 989 | 5311366 | ||
| BC130278.1 | - | 8 | 1228 | 40146309 | ||
| X77197.1 | - | 9 | 1322 | |||

CLCN4
Genomic Map