siRNA Details
| Gene Symbol: ACKR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001296.4 | NP_001287.2 | 3 | 1254 | ||
| GenBank mRNA | AK313561.1 | - | 3 | 1251 | ||
| BC008816.2 | - | 3 | 1242 | 3959017 | ||
| BC011631.2 | - | 3 | 1242 | 3958671 | ||
| BC020558.1 | - | 2 | 1173 | 3451472 | ||
| BC101629.1 | - | 1 | 1106 | 8069135 | ||
| BC112045.1 | - | 1 | 1106 | 8327513 | ||
| BT006800.1 | - | 1 | 1075 | |||
| Y12815.1 | - | 1 | 1087 | |||

ACKR2
Genomic Map