siRNA Details
| Gene Symbol: COL4A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000495.4 | NP_000486.1 | 41 | 4026 | ||
| NM_033380.2 | NP_203699.1 | 41 | 4026 | |||
| XM_011530849.2 | XP_011529151.2 | 42 | 3774 | |||
| XM_017029259.1 | XP_016884748.1 | 42 | 3774 | |||
| XM_017029260.1 | XP_016884749.1 | 42 | 3774 | |||
| XM_017029261.1 | XP_016884750.1 | 42 | 3774 | |||
| XM_017029262.1 | XP_016884751.1 | 42 | 3774 | |||
| XM_017029263.1 | XP_016884752.1 | 26 | 2852 | |||
| GenBank mRNA | BC151846.1 | - | 41 | 3988 | 8860442 | |
| KM282618.1 | - | 42 | 3999 | |||
| M31115.1 | - | 10 | 1005 | |||
| M58526.1 | - | 38 | 3502 | |||

COL4A5
Genomic Map