siRNA Details
| Gene Symbol: COX6B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001863.4 | NP_001854.1 | 2 | 279 | ||
| GenBank mRNA | AK312140.1 | - | 2 | 176 | ||
| BC001015.2 | - | 2 | 249 | 3344701 | ||
| BC002478.2 | - | 2 | 148 | 3347455 | ||
| BT006945.1 | - | 1 | 87 | |||
| CR456789.1 | - | 1 | 87 | |||
| CR542137.1 | - | 1 | 87 | |||
| X13923.1 | - | 2 | 131 | |||
| X54473.1 | - | 2 | 134 | |||

COX6B1
Genomic Map