siRNA Details
| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004375.4 | NP_004366.1 | 4 | 949 | ||
| NR_135677.1 | 4 | 949 | ||||
| XM_011524342.2 | XP_011522644.1 | 4 | 3188 | |||
| XM_017024192.1 | XP_016879681.1 | 4 | 3188 | |||
| XM_017024193.1 | XP_016879682.1 | 4 | 3188 | |||
| GenBank mRNA | AF044321.1 | - | 4 | 825 | ||
| AK293851.1 | - | 4 | 782 | |||
| BC005895.1 | - | 4 | 820 | 4282744 | ||
| CR541837.1 | - | 4 | 810 | |||
| U79270.1 | - | 3 | 440 | |||

COX11
Genomic Map