siRNA Details
| Gene Symbol: CPB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001871.2 | NP_001862.2 | 11 | 1262 | ||
| GenBank mRNA | AJ224866.1 | - | 11 | 1244 | ||
| AK313940.1 | - | 11 | 1250 | |||
| BC015338.1 | - | 11 | 1243 | 4422620 | ||
| BT009910.1 | - | 11 | 1239 | |||
| CR749369.1 | - | 2 | 3383 | |||
| M81057.1 | - | 11 | 1259 | |||

CPB1
Genomic Map