siRNA Details
| Gene Symbol: CREBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001310.3 | NP_001301.1 | 3 | 673 | ||
| GenBank mRNA | AB451461.1 | - | 2 | 333 | ||
| AF039081.1 | - | 3 | 609 | |||
| AK307202.1 | - | 4 | 556 | |||
| BC104873.1 | - | 3 | 517 | 8143876 | ||
| BC106052.1 | - | 3 | 614 | 6150812 | ||
| BC112157.1 | - | 3 | 517 | 8327625 | ||
| BY795169.2 | - | 3 | 673 | |||
| CR542250.1 | - | 2 | 333 | |||

CREBL2
Genomic Map