siRNA Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005206.4 | NP_005197.3 | 2 | 594 | ||
| NM_016823.3 | NP_058431.2 | 2 | 594 | |||
| GenBank mRNA | AK291060.1 | - | 2 | 559 | ||
| BC001718.1 | - | 2 | 546 | 3353237 | ||
| BC008506.1 | - | 2 | 577 | 3452137 | ||
| BC009837.1 | - | 2 | 559 | 4040536 | ||
| BT007277.1 | - | 2 | 444 | |||
| D10656.1 | - | 2 | 549 | |||
| KU177996.1 | - | 2 | 444 | |||
| KU177997.1 | - | 2 | 444 | |||

CRK
Genomic Map