siRNA Details
| Gene Symbol: CRYBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004076.4 | NP_004067.1 | 4, 5 | 405 | ||
| XM_011529902.2 | XP_011528204.1 | 4, 5 | 943 | |||
| GenBank mRNA | BC069479.1 | - | 4, 5 | 375 | 7262237 | |
| BM711382.1 | - | 4, 5 | 405 | |||

CRYBB3
Genomic Map