siRNA Details
| Gene Symbol: CRYGB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005210.3 | NP_005201.2 | 3 | 492 | ||
| XM_017003402.1 | XP_016858891.1 | 3 | 672 | |||
| GenBank mRNA | BC074944.2 | - | 3 | 492 | 30915273 | |
| BC074945.2 | - | 3 | 492 | 30915496 | ||
| BC117384.1 | - | 3 | 492 | 40125935 | ||
| BC117388.1 | - | 3 | 492 | 40125939 | ||
| BM666147.1 | - | 1 | 148 | |||
| BY794984.2 | - | 3 | 507 | |||
| KT326704.1 | - | 3 | 497 | |||

CRYGB
Genomic Map