siRNA Details
| Gene Symbol: CRYGS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017541.2 | NP_060011.1 | 3 | 430 | ||
| GenBank mRNA | AF161703.1 | - | 3 | 422 | ||
| AK314172.1 | - | 3 | 430 | |||
| AL133079.1 | - | 2 | 1280 | |||
| BC069478.1 | - | 3 | 428 | 7262271 | ||
| BC070241.1 | - | 4 | 718 | 30405628 | ||
| KT966393.1 | - | 3 | 418 | |||

CRYGS
Genomic Map