siRNA Details
| Gene Symbol: CSPG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001897.4 | NP_001888.2 | 9 | 5064 | ||
| GenBank mRNA | AY359468.1 | - | 9 | 4971 | ||
| X96753.1 | - | 9 | 4971 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CSPG4| Gene Symbol: CSPG4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001897.4 | NP_001888.2 | 9 | 5064 | ||
| GenBank mRNA | AY359468.1 | - | 9 | 4971 | ||
| X96753.1 | - | 9 | 4971 | |||