siRNA Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171171.1 | NP_001164642.1 | 2 | 813 | ||
| NM_001171172.1 | NP_001164643.1 | 2 | 773 | |||
| NM_001171174.1 | NP_001164645.1 | 2 | 709 | |||
| NM_001337.3 | NP_001328.1 | 2 | 665 | |||
| GenBank mRNA | AK225764.1 | - | 2 | 666 | ||
| AK307911.1 | - | 3 | 465 | |||
| AK312373.1 | - | 2 | 664 | |||
| BC028078.1 | - | 2 | 649 | 5216452 | ||
| U20350.1 | - | 2 | 660 | |||
| U28934.1 | - | 2 | 618 | |||

CX3CR1
Genomic Map