siRNA Details
| Gene Symbol: TEX28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205201.1 | NP_001192130.1 | 5 | 1076 | ||
| NM_001586.2 | NP_001577.1 | 5 | 1070 | |||
| XM_011531116.1 | XP_011529418.1 | 5 | 1114 | |||
| XM_011531117.1 | XP_011529419.1 | 6 | 1183 | |||
| XM_011531118.2 | XP_011529420.1 | 6 | 1311 | |||
| GenBank mRNA | BC026183.1 | - | 5 | 1081 | 5266650 | |
| U93720.1 | - | 5 | 1054 | |||
| Gene Symbol: OPN1MW | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000513.2 | NP_000504.1 | 6 | 1927 | ||
More Information
Set Membership: |
|

TEX28
Genomic Map