siRNA Details
Gene Symbol: TEX28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205201.1 | NP_001192130.1 | 3 | 300 | ||
NM_001586.2 | NP_001577.1 | 3 | 294 | |||
XM_011531116.1 | XP_011529418.1 | 3 | 338 | |||
XM_011531117.1 | XP_011529419.1 | 4 | 407 | |||
XM_011531118.2 | XP_011529420.1 | 4 | 535 | |||
GenBank mRNA | BC026183.1 | - | 3 | 305 | 5266650 | |
DB457033.1 | - | 3 | 302 | |||
DQ655913.2 | - | 4 | 372 | |||
U93720.1 | - | 3 | 278 |
