siRNA Details
| Gene Symbol: DAO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001917.4 | NP_001908.3 | 9 | 896 | ||
| XM_005268692.3 | XP_005268749.1 | 8 | 830 | |||
| XM_011538004.2 | XP_011536306.1 | 10 | 982 | |||
| XM_011538005.2 | XP_011536307.1 | 10 | 986 | |||
| GenBank mRNA | AK312995.1 | - | 9 | 883 | ||
| AX741655.1 | - | 9 | 886 | |||
| AX741656.1 | - | 9 | 886 | |||
| AX741658.1 | - | 8 | 771 | |||
| AX741659.1 | - | 6 | 658 | |||
| AX776478.1 | - | 9 | 886 | |||
| AX776479.1 | - | 9 | 886 | |||
| AX776480.1 | - | 11 | 1815 | |||
| AX786386.1 | - | 9 | 886 | |||
| BC029057.1 | - | 9 | 866 | 5184628 | ||
| BC074770.2 | - | 9 | 768 | 30915193 | ||
| BX538210.1 | - | 9 | 828 | |||
| KU178015.1 | - | 8 | 743 | |||

DAO
Genomic Map