siRNA Details
| Gene Symbol: DCC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005215.3 | NP_005206.2 | 15 | 2876 | ||
| XM_011525843.1 | XP_011524145.1 | 15 | 2876 | |||
| XM_011525844.2 | XP_011524146.1 | 12 | 1764 | |||
| XM_017025568.1 | XP_016881057.1 | 15 | 2876 | |||
| XM_017025569.1 | XP_016881058.1 | 15 | 2876 | |||
| XM_017025570.1 | XP_016881059.1 | 11 | 1772 | |||
| GenBank mRNA | AK302643.1 | - | 15 | 2181 | ||
| AK310385.1 | - | 12 | 1490 | |||
| BC036524.1 | - | 14 | 2062 | |||
| X76132.1 | - | 15 | 2260 | |||

DCC
Genomic Map