siRNA Details
| Gene Symbol: DMXL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290321.2 | NP_001277250.1 | 18 | 3913 | ||
| NM_001290322.2 | NP_001277251.1 | 18 | 5221 | |||
| NM_005509.5 | NP_005500.4 | 18 | 3913 | |||
| XM_005271909.4 | XP_005271966.1 | 19 | 3611 | |||
| XM_005271910.4 | XP_005271967.1 | 18 | 3956 | |||
| XM_005271912.2 | XP_005271969.1 | 19 | 3595 | |||
| XM_011543212.2 | XP_011541514.1 | 18 | 3956 | |||
| XM_011543213.2 | XP_011541515.1 | 19 | 3541 | |||
| XM_011543214.2 | XP_011541516.1 | 15 | 3122 | |||
| XM_011543215.2 | XP_011541517.1 | 14 | 2958 | |||
| XM_017009141.1 | XP_016864630.1 | 19 | 3611 | |||
| XM_017009142.1 | XP_016864631.1 | 19 | 3541 | |||
| XM_017009143.1 | XP_016864632.1 | 20 | 3790 | |||
| XM_017009144.1 | XP_016864633.1 | 18 | 3493 | |||
| XM_017009145.1 | XP_016864634.1 | 18 | 3363 | |||
| XM_017009146.1 | XP_016864635.1 | 18 | 4805 | |||
| XM_017009147.1 | XP_016864636.1 | 18 | 3956 | |||
| XM_017009148.1 | XP_016864637.1 | 17 | 3759 | |||
| GenBank mRNA | AB593142.1 | - | 18 | 5224 | ||
| AJ005821.1 | - | 18 | 3510 | |||
| BC144537.1 | - | 18 | 3456 | 9053064 | ||
| BC150616.1 | - | 18 | 3456 | 9056986 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001081371, AY590892.

DMXL1
Genomic Map