siRNA Details
| Gene Symbol: TIMM8A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004085.3 | NP_004076.1 | 2 | 542 | ||
| GenBank mRNA | AK312117.1 | - | 2 | 290 | ||
| BC006994.1 | - | 2 | 292 | 4051368 | ||
| BC015093.1 | - | 2 | 295 | 3684305 | ||
| BC070284.1 | - | 2 | 275 | 4281013 | ||
| CN410182.1 | - | 2 | 542 | |||
| U66035.1 | - | 2 | 272 | |||

TIMM8A
Genomic Map