siRNA Details
| Gene Symbol: DFNA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127453.1 | NP_001120925.1 | 5 | 742 | ||
| NM_001127454.1 | NP_001120926.1 | 4 | 757 | |||
| NM_004403.2 | NP_004394.1 | 5 | 987 | |||
| XM_017011802.1 | XP_016867291.1 | 4 | 513 | |||
| GenBank mRNA | AF007790.2 | - | 4 | 642 | ||
| AF073308.1 | - | 5 | 711 | |||
| AK094714.1 | - | 7 | 1315 | |||
| AK290775.1 | - | 5 | 718 | |||
| AK314402.1 | - | 5 | 727 | |||
| BC099911.1 | - | 5 | 703 | 5287362 | ||
| BC125065.1 | - | 5 | 709 | 40117291 | ||
| BC125066.1 | - | 4 | 479 | |||

DFNA5
Genomic Map