siRNA Details
Gene Symbol: DIO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000793.5 | NP_000784.2 | 3 | 476 | ||
NM_001007023.4 | NP_001007024.1 | 1 | 245 | |||
NM_001242502.2 | NP_001229431.1 | 1 | 245 | |||
NM_001242503.2 | NP_001229432.1 | 1 | 245 | |||
NM_001324462.1 | NP_001311391.1 | 2 | 492 | |||
NM_013989.4 | NP_054644.1 | 1 | 245 | |||
GenBank mRNA | AB041843.1 | - | 1 | 118 | ||
AF093774.1 | - | 1 | 245 | |||
AK292210.1 | - | 3 | 476 | |||
BC063118.1 | - | 1 | 128 | |||
BC074882.2 | - | 1 | 121 | 30915240 | ||
BC136514.1 | - | 1 | 115 | 9020504 | ||
CD691767.1 | - | 1 | 178 | |||
JK546617.1 | - | 2 | 403 | |||
U53506.1 | - | 1 | 224 |
