siRNA Details
| Gene Symbol: DPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001937.4 | NP_001928.2 | 3 | 505 | ||
| GenBank mRNA | BC033736.1 | - | 3 | 491 | 5176855 | |
| BP257325.1 | - | 3 | 505 | |||
| Z22865.1 | - | 3 | 487 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DPT| Gene Symbol: DPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001937.4 | NP_001928.2 | 3 | 505 | ||
| GenBank mRNA | BC033736.1 | - | 3 | 491 | 5176855 | |
| BP257325.1 | - | 3 | 505 | |||
| Z22865.1 | - | 3 | 487 | |||