siRNA Details
| Gene Symbol: DRG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001388.4 | NP_001379.1 | 3 | 435 | ||
| XM_005256499.2 | XP_005256556.1 | 3 | 417 | |||
| XM_005256500.2 | XP_005256557.1 | 3 | 412 | |||
| XM_011523704.1 | XP_011522006.1 | 3 | 417 | |||
| XM_011523705.1 | XP_011522007.1 | 3 | 417 | |||
| XM_011523706.1 | XP_011522008.1 | 3 | 330 | |||
| XM_011523707.1 | XP_011522009.1 | 3 | 376 | |||
| XM_017024296.1 | XP_016879785.1 | 3 | 330 | |||
| XM_017024297.1 | XP_016879786.1 | 3 | 376 | |||
| GenBank mRNA | AB209340.1 | - | 1 | 2602 | ||
| AK295575.1 | - | 3 | 323 | |||
| AK297569.1 | - | 3 | 325 | |||
| AK313362.1 | - | 3 | 354 | |||
| BC000493.2 | - | 3 | 333 | 2820542 | ||
| BT006976.1 | - | 3 | 278 | |||
| DC298961.1 | - | 3 | 430 | |||
| X80754.1 | - | 3 | 325 | |||

DRG2
Genomic Map