siRNA Details
| Gene Symbol: DSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004949.4 | NP_004940.1 | 7, 8 | 1393 | ||
| NM_024422.4 | NP_077740.1 | 7, 8 | 1393 | |||
| XM_005258206.4 | XP_005258263.1 | 7, 8 | 894 | |||
| GenBank mRNA | AK290313.1 | - | 7, 8 | 1084 | ||
| AK297029.1 | - | 8, 9 | 1059 | |||
| BC063291.1 | - | 7, 8 | 1375 | 30337780 | ||
| BX648425.1 | - | 7, 8 | 945 | |||
| CR749226.1 | - | 7, 8 | 1112 | |||
| EF017811.1 | - | 7, 8 | 932 | |||
| EF017812.1 | - | 7, 8 | 932 | |||
| X56807.1 | - | 7, 8 | 972 | |||

DSC2
Genomic Map