siRNA Details
| Gene Symbol: DSC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004949.4 | NP_004940.1 | 10 | 1874 | ||
| NM_024422.4 | NP_077740.1 | 10 | 1874 | |||
| XM_005258206.4 | XP_005258263.1 | 10 | 1375 | |||
| GenBank mRNA | AK290313.1 | - | 10 | 1565 | ||
| AK297029.1 | - | 11 | 1534 | |||
| BC063291.1 | - | 10 | 1856 | 30337780 | ||
| BX648425.1 | - | 10 | 1426 | |||
| CR749226.1 | - | 10 | 1593 | |||
| EF017811.1 | - | 10 | 1413 | |||
| EF017812.1 | - | 10 | 1413 | |||
| X56807.1 | - | 10 | 1453 | |||

DSC2
Genomic Map