siRNA Details
| Gene Symbol: DSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001942.3 | NP_001933.2 | 15 | 2578 | ||
| GenBank mRNA | AF097935.1 | - | 15 | 2579 | ||
| AK302888.1 | - | 4 | 752 | |||
| X56654.1 | - | 15 | 2443 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DSG1| Gene Symbol: DSG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001942.3 | NP_001933.2 | 15 | 2578 | ||
| GenBank mRNA | AF097935.1 | - | 15 | 2579 | ||
| AK302888.1 | - | 4 | 752 | |||
| X56654.1 | - | 15 | 2443 | |||