siRNA Details
| Gene Symbol: DUSP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004419.3 | NP_004410.3 | 4 | 1077 | ||
| GenBank mRNA | AB209823.1 | - | 4 | 1023 | ||
| BC062545.1 | - | 4 | 994 | 5808876 | ||
| U15932.2 | - | 4 | 1029 | |||
| U16996.1 | - | 4 | 868 | |||

DUSP5
Genomic Map