siRNA Details
| Gene Symbol: DYRK1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001396.3 | NP_001387.2 | 4, 5 | 595 | ||
| NM_101395.2 | NP_567824.1 | 6, 7 | 769 | |||
| NM_130436.2 | NP_569120.1 | 4, 5 | 1963 | |||
| NM_130438.2 | NP_569122.1 | 4, 5 | 596 | |||
| XM_006723976.3 | XP_006724039.1 | 5, 6 | 667 | |||
| XM_006723977.3 | XP_006724040.1 | 5, 6 | 992 | |||
| XM_006723978.3 | XP_006724041.1 | 5, 6 | 1721 | |||
| XM_006723979.3 | XP_006724042.1 | 5, 6 | 827 | |||
| XM_011529482.2 | XP_011527784.1 | 4, 5 | 689 | |||
| XM_011529483.2 | XP_011527785.1 | 5, 6 | 3535 | |||
| XM_011529484.2 | XP_011527786.1 | 4, 5 | 662 | |||
| XM_011529485.2 | XP_011527787.1 | 4, 5 | 1178 | |||
| XM_017028284.1 | XP_016883773.1 | 5, 6 | 626 | |||
| XM_017028285.1 | XP_016883774.1 | 5, 6 | 872 | |||
| XM_017028286.1 | XP_016883775.1 | 7, 8 | 869 | |||
| GenBank mRNA | AF108830.1 | - | 6, 7 | 769 | ||
| D85759.1 | - | 4, 5 | 561 | |||
| D86550.1 | - | 4, 5 | 1966 | |||
| U52373.1 | - | 4, 5 | 595 | |||

DYRK1A
Genomic Map