siRNA Details
| Gene Symbol: EDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001302269.1 | NP_001289198.1 | 2 | 262 | ||
| NM_001956.4 | NP_001947.1 | 2 | 262 | |||
| NR_126098.1 | 2 | 171 | ||||
| XM_017000512.1 | XP_016856001.1 | 2 | 277 | |||
| GenBank mRNA | BC034393.1 | - | 2 | 205 | 5185394 | |
| BI761655.1 | - | 2 | 161 | |||
| HY177176.1 | - | 2 | 263 | |||
| M65199.1 | - | 2 | 250 | |||
| X55177.1 | - | 2 | 238 | |||

EDN2
Genomic Map