siRNA Details
| Gene Symbol: EDN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001956.4 | NP_001947.1 | 4, 5 | 530 | ||
| NR_126098.1 | 4, 5 | 439 | ||||
| GenBank mRNA | BC034393.1 | - | 4, 5 | 473 | 5185394 | |
| BI761655.1 | - | 4, 5 | 428 | |||
| M65199.1 | - | 4, 5 | 518 | |||
| S63835.1 | - | 2, 3 | 143 | |||
| S63837.1 | - | 3, 4 | 266 | |||
| S63838.1 | - | 4, 5 | 318 | |||
| X55177.1 | - | 4, 5 | 506 | |||

EDN2
Genomic Map